Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BATF Antibody (687706), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311043 Supplier Novus Biologicals Supplier No. FAB8054PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BATF Monoclonal specifically detects BATF in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BATF
Applications Flow Cytometry
Classification Monoclonal
Clone 687706
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein
Host Species Mouse
Immunogen E. coli-derived recombinant human BATF , Met1-Pro125, Accession # Q16520
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10538
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.