Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Batimastat
GREENER_CHOICESDP

Catalog No. 29611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-611 1 mg
29-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-611 Supplier Tocris Bioscience™ Supplier No. 2961/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, broad spectrum MMP inhibitor

Potent, broad spectrum MMP inhibitor (IC50 values are 3, 4, 4, 6 and 20 nM for MMP -1, -2, -9, -7 and -3 respectively). Exhibits antiproliferative, anti-invasive and antimetastatic activity in human ovarian carcinoma xenografts in vivo.

Chemical Identifiers

CAS 130370-60-4
Molecular Formula C23H31N3O4S2

Specifications

Quantity 1 mg
Formula Weight 477.64
Purity >99%
Molecular Weight (g/mol) 477.638
Chemical Name or Material Batimastat
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.