Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Antibody, Novus Biologicals™
SDP

Catalog No. NB10056096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056096 0.05 mL
NB10056096T 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056096 Supplier Novus Biologicals Supplier No. NB10056096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Bax Polyclonal specifically detects Bax in Canine, Gerbil, Human, Mouse, Rat samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Bax
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. Q07813
Gene Alias apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4
Gene Symbols BAX
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 43-60 (PELTLEQPPQDASTKKLS) of mouse Bax alpha was used as immunogen, GenBank no. NP_031553.1.
Molecular Weight of Antigen 21 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 581
Test Specificity Bax
Target Species Canine, Mouse, Gerbil, Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.