Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bax channel blocker
GREENER_CHOICE

Catalog No. 216010R
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 21-601-0R

Supplier: Tocris Bioscience™ 2160/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Allosteric inhibitor of Bax channel activation

Bax channel blocker is a allosteric inhibitor of Bax channel activation. Binds inactive Bax at allosteric site and inhibits tBID-mediated Bax activation (IC50 = 3.3 μM). Selectively inhibits Bax-dependent apoptosis. Potent inhibitor of Bax-mediated mitochondrial cytochrome c release (IC50 = 0.52 μM).

Specifications

Quantity 10 mg
Product Type Bax channel blocker
Purity >98%
Molecular Weight (g/mol) 540.12
Inhibitors Bax channel blocker

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.