Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax inhibitor 1 Antibody (20F430), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP224919W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224919W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224919W Supplier Novus Biologicals Supplier No. NBP224919AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Bax inhibitor 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 20F430
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. P55061
Gene Alias BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6
Gene Symbols TMBIM6
Host Species Mouse
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal, internal and C-terminal regions of human B1. Epitope mapping indicates that the antibody recognizes an epitope in the N-terminal region of Bi-1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 7009
Test Specificity This antibody is reactive to Human, Mouse, Rat.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.