Learn More
Bax inhibitor 1 Antibody (20F565), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Bax inhibitor 1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 20F565 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6 |
| Gene Symbols | TMBIM6 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.