Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Mouse anti-Human, Monkey, DyLight 680, Clone: 2D2, Novus Biologicals™
SDP

Catalog No. NB019513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019513 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019513 Supplier Novus Biologicals Supplier No. NBP233092FR100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bax Monoclonal specifically detects Bax in Human, Monkey, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Bax
Applications ELISA
Classification Monoclonal
Clone 2D2
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4
Host Species Mouse
Immunogen A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 581
Target Species Human, Monkey, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.