Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 1797
GREENER_CHOICE

Catalog No. p-7249488
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective P2X4 antagonist

BAY 1797 is a selective purinergic P2X4 receptor antagonist (IC50 values are 108, 112 and 233 nM for the human, mouse and rat P2X4, respectively). It is selective for P2X4 over P2X3, P2X7, and P2X1 receptors (IC50 values are 8.3, 10.6 and >50 μM, respectively, for the human receptors). BAY 1797 dose-dependently reduces prostaglandin E2 levels and exhibits anti-inflammatory and analgesic effects in an in vivo model of inflammatory pain. Orally active.

Specifications

Chemical Name or Material N-[3-(Aminosulfonyl)-4-(3-chlorophenoxy)phenyl]benzeneacetamide
CAS 2055602-83-8
Quantity 10 mg
Target P2X4
Molecular Formula C20H17ClN2O4S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.