Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 41-2272
GREENER_CHOICE

Catalog No. p-4816928
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Soluble guanylyl cyclase (sGC) activator

Activator of soluble guanylyl cyclase (sGC); acts at a nitric oxide (NO)-independent regulatory site in the sGC α1 subunit. Inhibits platelet aggregation (IC50 = 36 nM) and phenylephrine-induced contractions of rabbit aorta (IC50 = 0.30 μM). Also reduces vascular smooth muscle growth through cAMP- and cGMP-dependent PKA and PKG pathways.

Chemical Identifiers

CAS 256376-24-6
Molecular Formula C20H17FN6

Specifications

Quantity 10 mg
Formula Weight 360.39
Purity >98%
Molecular Weight (g/mol) 360.40
Chemical Name or Material BAY 41-2272
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.