Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BAY 707

Click to view available options
Quantity:
5 mg
25 mg
Description
BAY 707 is a potent and selective MTH1 inhibitor (IC50 = 2.3 nM), whicheExhibits no significant activity at 1 μM concentration against a panel of 97 kinases. BAY 707 is cell-permeable and active in vivo, and displays no antitumor activity in vitro or in vivo.
Specifications
Specifications
Chemical Name or Material | N-Ethyl-4-[(3 S)-3-methyl-4-morpholinyl]-1 H-pyrrolo[2,3-b]pyridine-2-carboxamide |
CAS | 2109805-96-9 |
Quantity | 25 mg |
Target | MTH1 Inhibitors |
Molecular Formula | C15H20N4O2 |
Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction