Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ BAY 8002
Click to view available options
Quantity:
10 mg
50 mg
Description
BAY 8002 is a potent dual MCT1/2 inhibitor (IC50 values are 3, 8, and 12 nM at rat (C6), human (DLD-1) and mouse (4T1) MCT1 respectively; IC50 values of human MCT1, MCT2, and MCT4 isoforms recombinantly expressed in X. laevis oocytes are 1, 5 and >500 nM, respectively); suppresses bidirectional lactate transport. Inhibits cellular SNARF-5 fluorescence change (IC50 = 85 nM) in MCT1-expressing DLD-1 cells. Arrests growth and increases intratumor lactate levels of Raji tumor xenografts in mice. Orally bioavailable.
Specifications
Specifications
Quantity | 10 mg |
Molecular Formula | C20H14ClNO5S |
Formula Weight | 415.85 |
Product Type | Inhibitor |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction