Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAZ2A Antibody, Novus Biologicals™
SDP

Catalog No. NB394786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB394786 25 μL
NBP255854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB394786 Supplier Novus Biologicals Supplier No. NBP25585425UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

BAZ2A Polyclonal specifically detects BAZ2A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BAZ2A
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias bromodomain adjacent to zinc finger domain protein 2A, bromodomain adjacent to zinc finger domain, 2A, DKFZp781B109, FLJ45876, hWALp3, KIAA0314FLJ13768, Tip5, TIP5FLJ13780, Transcription termination factor I-interacting protein 5, TTF-I interacting peptide 5, TTF-I-interacting protein 5, WALp3
Gene Symbols BAZ2A
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SLEVPLTASVTSPKASPVTSPAAAFPTASPANKDVSSFLETTADVEEITGEGLTASGSGDVMRRRIATPEEVRLPLQHGWRREVRIKKGSHR
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11176
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.