Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BB515 Mouse Anti-Human Integrin α1 (CD49a), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570157
Description
Integrin α1 (CD49a) Monoclonal antibody specifically detects Integrin α1 (CD49a) in Human samples. It is validated for Flow cytometry
Specifications
Integrin α1 (CD49a) | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
ITGA1; CD49a; VLA-1; VLA1; integrin alpha-1 | |
Human HLA-DR CTL line | |
25 Tests | |
3672 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
TS2/7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction