Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BB515 Mouse Anti-Human Integrin α1 (CD49a), BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | Integrin α1 (CD49a) |
---|---|
Clone | TS2/7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 515 (BB515) |
Description
Integrin α1 (CD49a) Monoclonal antibody specifically detects Integrin α1 (CD49a) in Human samples. It is validated for Flow cytometrySpecifications
Integrin α1 (CD49a) | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
Mouse | |
Human | |
ITGA1; CD49a; VLA-1; VLA1; integrin alpha-1 | |
Human HLA-DR CTL line | |
Affinity Chromatography |
TS2/7 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3672 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title