Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BB515 Mouse Anti-Integrin α7 (ITGA7), BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | Integrin α7 (ITGA7) |
---|---|
Clone | 3C12 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Integrin α7 (ITGA7) Monoclonal antibody specifically detects Integrin α7 (ITGA7) in Mouse|Human samples. It is validated for Flow cytometrySpecifications
Integrin α7 (ITGA7) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Myoblasts | |
Affinity Chromatography |
3C12 | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
Mouse | |
Mouse, Human | |
ITGA7; Integrin α7; integrin alpha-7; α7 Integrin | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title