Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BB515 Mouse Anti-Mouse CD369 (CLEC7A), BD Horizon™
Mouse Monoclonal Antibody
$140.00 - $326.00
Specifications
Antigen | CD369 (CLEC7A) |
---|---|
Clone | 2F3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD369 (CLEC7A) Monoclonal antibody specifically detects CD369 (CLEC7A) in Mouse samples. It is validated for Flow cytometrySpecifications
CD369 (CLEC7A) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
56644 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2F3 | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
Mouse | |
Mouse | |
Clec7a; CD369; BGR; Beta-glucan receptor; Bgr; Dectin-1; Dectin1; dectin-1 | |
Mouse Dectin-1 Recombinant Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title