Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Sabouraud Dextrose Agar

Catalog No. B11584
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

For cultivation of yeasts, molds and aciduric microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type Sabouraud Dextrose Agar
Product Type Agar
Formulation Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L
For Use With (Application) Cultivation of Yeasts, molds and aciduric microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.