Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBS2 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB136047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136047 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136047 Supplier Novus Biologicals Supplier No. NBP306335H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BBS2 Polyclonal specifically detects BBS2 in Human samples. It is validated for Western Blot.

Specifications

Antigen BBS2
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias Bardet-Biedl syndrome 2, Bardet-Biedl syndrome 2 protein, BBS, MGC20703
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human BBS2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 583
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.