Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBS5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096062 20 μL
NB096061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096062 Supplier Novus Biologicals Supplier No. NBP30469120UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BBS5 Polyclonal specifically detects BBS5 in Human samples. It is validated for Western Blot.

Specifications

Antigen BBS5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Bardet-Biedl syndrome 5, Bardet-Biedl syndrome 5 protein, DKFZp762I194
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-341 of human BBS5 (NP_689597.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 129880
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.