Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCA-225 (BRST-1) Mouse anti-Human, Unconjugated, Clone: CU18, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF219825UL
Description
BCA-225 (BRST-1) Monoclonal antibody specifically detects BCA-225 (BRST-1) in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
BCA-225 (BRST-1) | |
Monoclonal | |
Unconjugated | |
Mouse | |
225-250 kDa calculated. | |
25 μL | |
Primary | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
CU18 | |
Breast Carcinoma Associated Glycoprotein;Breast cancer antigen 225;BRST-1 | |
BCA-225 protein secreted by the T47D (Clone 11) human breast carcinoma cell line. | |
Protein G Purified | |
Apoptosis and Cancer | |
Clone CU18 is a mouse monoclonal antibody that detects human breast carcinoma associated glycoprotein BCA-225. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction