Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCA-225 (BRST-1) Mouse anti-Human, Unconjugated, Clone: CU18, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | BCA-225 (BRST-1) |
---|---|
Clone | CU18 |
Applications | ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF21982MI
|
MilliporeSigma
MABF219825UL |
25 μL |
Each for $189.29
|
|
|||||
MABF21981MI
|
MilliporeSigma
MABF2198100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
BCA-225 (BRST-1) Monoclonal antibody specifically detects BCA-225 (BRST-1) in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.Specifications
BCA-225 (BRST-1) | |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
BCA-225 protein secreted by the T47D (Clone 11) human breast carcinoma cell line. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
225-250 kDa calculated. |
CU18 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
Breast Carcinoma Associated Glycoprotein;Breast cancer antigen 225;BRST-1 | |
IgG1 κ | |
Protein G Purified | |
Clone CU18 is a mouse monoclonal antibody that detects human breast carcinoma associated glycoprotein BCA-225. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title