Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCA3 Antibody (OTI1E3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272057IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272057IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272057IR Supplier Novus Biologicals Supplier No. NBP272057IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCA3 Monoclonal specifically detects BCA3 in Human samples. It is validated for Western Blot.

Specifications

Antigen BCA3
Applications Western Blot
Classification Monoclonal
Clone OTI1E3
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AKIP1koyt binding protein 2, BCA3koyt binding protein 3, breast cancer associated gene 3, Breast cancer-associated gene 3 protein, chromosome 11 open reading frame 17, koyt binding protein 1, PKA-interacting protein, proline-rich protein BCA3, protein kinase A-interacting protein 1
Gene Symbols AKIP1
Host Species Mouse
Immunogen Full length recombinant protein of human BCA3 (NP_878904) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 56672.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.