Learn More
Description
Specifications
Specifications
| Antigen | BCAM/CD239 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2 |
| Conjugate | FITC |
| Dilution | ELISA, Sandwich ELISA Detection |
| Formulation | PBS |
| Gene Alias | antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human BCAM/CD239 (NP_005572.2; Met1-Ala547). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
