Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAM/CD239 Antibody (002), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB066435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB066435 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB066435 Supplier Novus Biologicals Supplier No. NBP289356JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen BCAM/CD239
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Janelia Fluor 549
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human BCAM/CD239 (NP_005572.2; Met1-Ala547).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4059
Test Specificity No cross-reactivity in ELISA with: Human ALCAM Human CD31 Human EpCAM Human ICAM1 Human ICAM2 Human ICAM3 Human MCAM Human VCAM1
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.