Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAM/CD239 Antibody (87207), Biotin, Novus Biologicals™
SDP

Catalog No. NB304887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304887 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304887 Supplier Novus Biologicals Supplier No. FAB1481B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry, Western Blot.

Specifications

Antigen BCAM/CD239
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry, Western Blot
Classification Monoclonal
Clone 87207
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready
Formulation PBS
Gene Alias antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BCAM, Glu32-Ala547, Accession # CAA58449
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4059
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.