Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAM/CD239 Antibody (87207), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB175216 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175216 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175216 Supplier Novus Biologicals Supplier No. FAB1481J
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCAM/CD239 Monoclonal antibody specifically detects BCAM/CD239 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)

Specifications

Antigen BCAM/CD239
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 87207
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BCAM, Glu32-Ala547, Accession # CAA58449
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4059
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.