Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAM/CD239 Antibody (MM0107-1M39), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB361398 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361398 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361398 Supplier Novus Biologicals Supplier No. NBP212050MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCAM/CD239 Monoclonal antibody specifically detects BCAM/CD239 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen BCAM/CD239
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0107-1M39
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19
Host Species Mouse
Immunogen Human recombinant BCAM/CD239 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, CD Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4059
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.