Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM (CD239) Mouse anti-Human, Clone: F241, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1940100UG
Description
BCAM (CD239) Monoclonal antibody specifically detects BCAM (CD239) in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blotting.
Specifications
BCAM (CD239) | |
Monoclonal | |
1 mg/mL | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
K562 cells transfected with full-length BCAM protein. | |
100 μg | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
F241 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction