Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM (CD239) Mouse anti-Human, Clone: F241, MilliporeSigma™
Mouse Monoclonal Antibody
$191.34 - $481.79
Specifications
Antigen | BCAM (CD239) |
---|---|
Clone | F241 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS194025
|
MilliporeSigma
MABS194025UG |
25 μg |
Each for $191.34
|
|
|||||
MABS1940100
|
MilliporeSigma
MABS1940100UG |
100 μg |
Each for $481.79
|
|
|||||
Description
BCAM (CD239) Monoclonal antibody specifically detects BCAM (CD239) in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blotting.Specifications
BCAM (CD239) | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
F241 | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
K562 cells transfected with full-length BCAM protein. | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title