Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAP/PIK3AP1 Antibody (OTI7A11), HRP, Novus Biologicals™
SDP

Catalog No. NBP272368H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272368H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272368H Supplier Novus Biologicals Supplier No. NBP272368H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCAP/PIK3AP1 Monoclonal specifically detects BCAP/PIK3AP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen BCAP/PIK3AP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7A11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias 1810044J04Rik, B cell adaptor protein, BCAPB-cell adapter for phosphoinositide 3-kinase, B-cell phosphoinositide 3-kinase adapter protein 1, FLJ35564, phosphoinositide 3-kinase adapter protein 1, phosphoinositide-3-kinase adaptor protein 1, RP11-34E5.3
Gene Symbols PIK3AP1
Host Species Mouse
Immunogen Full length recombinant protein of human BCAP/PIK3AP1 (NP_689552) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 118788.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.