Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAP31 Antibody (12G12), Novus Biologicals™
SDP

Catalog No. NB298015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298015 100 μL
NB298014 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298015 Supplier Novus Biologicals Supplier No. NBP326481100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BCAP31 Monoclonal antibody specifically detects BCAP31 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen BCAP31
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 12G12
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Bap31,6C6-AG, BAP31DXS1357ECDM, B-cell receptor-associated protein 31, BCR-associated protein 31, BCR-associated protein Bap31,6C6-AG tumor-associated antigen, p28, p28 Bap31,6C6-Ag, Protein CDM
Host Species Rabbit
Immunogen A synthesized peptide derived from Human BCAP31 [UniProt P51572]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10134
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.