Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAP31 Antibody (JG37-81), Novus Biologicals™
SDP

Catalog No. NBP275416 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275416 Supplier Novus Biologicals Supplier No. NBP275416

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

BCAP31 Monoclonal specifically detects BCAP31 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen BCAP31
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JG37-81
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias Bap31,6C6-AG, BAP31DXS1357ECDM, B-cell receptor-associated protein 31, BCR-associated protein 31, BCR-associated protein Bap31,6C6-AG tumor-associated antigen, p28, p28 Bap31,6C6-Ag, Protein CDM
Gene Symbols BCAP31
Host Species Rabbit
Immunogen Recombinant protein within Human BCAP31 aa 81-220/ 246. (SwissProt: P51572 Human; SwissProt: Q61335 Mouse)
Molecular Weight of Antigen 28 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Apoptosis, Mitochondrial Mediated Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10134.0
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.