Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAR3 Antibody, Novus Biologicals™
SDP

Catalog No. NB300825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300825 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300825 Supplier Novus Biologicals Supplier No. NB300825
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BCAR3 Polyclonal specifically detects BCAR3 in Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen BCAR3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_003558.
Gene Alias breast cancer estrogen resistance 3, dJ1033H22.2 (breast cancer estrogen resistance 3), KIAA0554, Novel SH2-containing protein 2, NSP2SH2D3Bbreast cancer estrogen resistance protein 3, SH2 domain-containing protein 3B
Gene Symbols BCAR3
Host Species Goat
Immunogen Peptide with sequence C-RKLEPPPVKQAEL corresponding to C-Terminus according to NP_003558.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8412
Test Specificity Cross reacts with Mouse. Expected from sequence similarity: Human, Dog, Pig and Cow.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.