Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAT1 Antibody (OTI3F5), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N270242J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270242J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270242J549 Supplier Novus Biologicals Supplier No. NBP270242JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCAT1 Monoclonal specifically detects BCAT1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen BCAT1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3F5
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39
Gene Symbols BCAT1
Host Species Mouse
Immunogen Full length recombinant protein of human BCAT1 (NP_005495) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 586.0
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.