Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCCIP Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB091975 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB091975 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB091975 Supplier Novus Biologicals Supplier No. NBP300296G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCCIP Polyclonal specifically detects BCCIP in Human samples. It is validated for ELISA.

Specifications

Antigen BCCIP
Applications ELISA
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias BCCIPalpha, BRCA2 and CDKN1A interacting protein, BRCA2 and CDKN1A-interacting protein, BRCA2 and Cip1/p21 interacting protein, cdk inhibitor p21 binding protein, P21- and CDK-associated protein 1, Protein TOK-1, TOK-1, TOK-1alpha, TOK1BCCIPbeta, TOK-1beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant BCCIP (AAH09771.1; Met1-Val314).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56647
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.