Learn More
Description
Specifications
Specifications
| Antigen | Bcl-10 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | BL10/411 |
| Conjugate | Alexa Fluor 647 |
| Dilution | ELISA |
| Gene Alias | B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB |
| Gene Symbols | BCL10 |
| Host Species | Mouse |
| Immunogen | Human Bcl-10 recombinant protein (epitope aa122-168) (Uniprot: O95999) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
