Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-10 Mouse anti-Human, Clone: rBL10/411, Novus Biologicals™
SDP

Catalog No. NB122412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122412 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122412 Supplier Novus Biologicals Supplier No. NBP30746920UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-10 Monoclonal specifically detects Bcl-10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.

Specifications

Antigen Bcl-10
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rBL10/411
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB
Host Species Mouse
Immunogen Human Bcl-10 recombinant protein (epitope aa122-168)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8915
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.