Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Antibody (BCL2/6426R), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB362976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362976 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362976 Supplier Novus Biologicals Supplier No. NBP308574MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Bcl-2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BCL2/6426R
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Host Species Rabbit
Immunogen A synthetic peptide, aa41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein. (Uniprot: P10415 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Mitophagy, Oncogenes, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.