Learn More
Description
Specifications
Specifications
| Antigen | Bcl-2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 |
| Gene Symbols | BCL2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 67-86 (ARTSPLRPLVATAGPALSPV) of mouse and rat Bcl-2 was used as immunogen; GenBank no gb/AAP47159.1. This sequence is 100% conserved in mouse and rat Bcl-2 alpha (236 amino acids) and beta (100 amino acids) isoforms. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
