Learn More
Description
Specifications
Specifications
| Antigen | Bcl-2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 |
| Gene Symbols | BCL2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to amino acids 61-76 (ASRDPVARTSPLQTPAC) of human Bcl-2 was used as immunogen; GenBank no NP_000648.2. This sequence is 100% conserved in human Bcl-2 alpha (239 amino acids) and beta (205 amino acids) isoforms. The literature indicates that the immunogen sequence is not conserved between human Bcl-2 and other species. Therefore this antibody may be particularly useful for detecting human Bcl-2 in transgenic studies. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
