Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $328.50

Specifications

Antigen Bcl-2
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056102T
SDP
View Documents
Novus Biologicals
NB10056102SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB10056102
SDP
View Documents
Novus Biologicals
NB10056102
0.05 mL
Each for $328.50
Only null left
Add to Cart
 
Description

Description

Bcl-2 Polyclonal specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Bcl-2
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Rabbit
Apoptosis, Autophagy, Cancer, Tumor Suppressors
Whole antisera with 0.05% Sodium Azide
596
A synthetic peptide corresponding to amino acids 61-76 (ASRDPVARTSPLQTPAC) of human Bcl-2 was used as immunogen; GenBank no NP_000648.2. This sequence is 100% conserved in human Bcl-2 alpha (239 amino acids) and beta (205 amino acids) isoforms. The literature indicates that the immunogen sequence is not conserved between human Bcl-2 and other species. Therefore this antibody may be particularly useful for detecting human Bcl-2 in transgenic studies.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen
Polyclonal
Antisera
RUO
Human
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
BCL2
IgG
Unpurified
Bcl-2
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.