Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse anti-Human, DyLight 550, Clone: 100/D5, Novus Biologicals™
SDP

Catalog No. NB019628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019628 Supplier Novus Biologicals Supplier No. NBP233315R100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-2 Monoclonal specifically detects Bcl-2 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Bcl-2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 100/D5
Conjugate DyLight 550
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1-5ug/ml for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation 50mM Sodium Borate
Gene Alias apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Host Species Mouse
Immunogen The synthetic peptide corresponding to amino acids range from 41-64 (GAAPAPGIFSSQPG-Cys), of human Bcl-2 was used as immunogen to generate the antibody; GenBank no. NP_000648.2 (Pezzella et al, 1990).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cellular Markers, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.