Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 related protein A1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357539 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357539 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357539 Supplier Novus Biologicals Supplier No. NBP176715MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bcl-2 related protein A1 Polyclonal antibody specifically detects Bcl-2 related protein A1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Bcl-2 related protein A1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Bfl-1. The immunogen is located within the last 50 amino acids of Bfl-1. Amino Acid Squence: ENGFVKKFEPKSG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 597
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.