Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 related protein A1 Antibody (OTI3D10), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N270246A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270246A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270246A647 Supplier Novus Biologicals Supplier No. NBP270246AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-2 related protein A1 Monoclonal specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Bcl-2 related protein A1
Applications Western Blot
Classification Monoclonal
Clone OTI3D10
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS
Gene Symbols BCL2A1
Host Species Mouse
Immunogen Full length recombinant protein of human Bcl-2 related protein A1 (NP_004040) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 597.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.