Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 related protein A1 Antibody (OTI3D10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP270246 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270246 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270246 Supplier Novus Biologicals Supplier No. NBP270246
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-2 related protein A1 Monoclonal specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Bcl-2 related protein A1
Applications Western Blot
Classification Monoclonal
Clone OTI3D10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS
Gene Symbols BCL2A1
Host Species Mouse
Immunogen Full length human recombinant protein of human BCL2A1 (NP_004040) produced in E.coli.
Molecular Weight of Antigen 20 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 597
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.