Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bcl-3 Monoclonal Antibody (Ham 150-3.5), FITC
Armenian Hamster Monoclonal Antibody
$317.50 - $566.50
Specifications
Antigen | Bcl-3 |
---|---|
Clone | Ham 150-3.5 |
Concentration | 0.1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517667
|
Invitrogen™
MA517667 |
100 μg |
Each for $317.50
|
|
|||||
PIMA517668
|
Invitrogen™
MA517668 |
300 μg |
Each for $566.50
|
|
|||||
Description
Bcl-3 Monoclonal Antibody for Flow
B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.Specifications
Bcl-3 | |
0.1 mg/mL | |
Flow Cytometry | |
FITC | |
Armenian Hamster | |
RUO | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Q9Z2F6 | |
12051 | |
recombinant Bcl-3-6-histidine tag. | |
Antibody |
Ham 150-3.5 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Monoclonal | |
Liquid | |
IgG | |
Mouse | |
Bcl3 | |
AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3 | |
Bcl3 | |
Primary | |
Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title