Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-6 Antibody (SPM602), PerCP, Novus Biologicals™
SDP

Catalog No. NB384066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384066 Supplier Novus Biologicals Supplier No. NBP308851PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Bcl-6
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM602
Conjugate PerCP
Formulation PBS
Gene Alias B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S
Host Species Mouse
Immunogen Recombinant human bcl-6 protein (Uniprot: P41182)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.