Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl G Antibody (OTI6D1), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272204A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272204A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272204A94 Supplier Novus Biologicals Supplier No. NBP272204AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl G Monoclonal specifically detects Bcl G in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Bcl G
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6D1
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias apoptosis facilitator Bcl-2-like protein 14, Apoptosis regulator Bcl-G, Bcl2-L-14, BCL2-like 14 (apoptosis facilitator), BCL-G, BCLGbcl2-L-14
Gene Symbols BCL2L14
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 116-216 of human Bcl G (NP_620048) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79370.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.