Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL-W/BCL2L2 Antibody (CPTC-BCL2L2-2), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB384509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384509 Supplier Novus Biologicals Supplier No. NBP308778AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCL-W/BCL2L2 Monoclonal antibody specifically detects BCL-W/BCL2L2 in Human samples. It is validated for Western Blot, Protein Array

Specifications

Antigen BCL-W/BCL2L2
Applications Western Blot, Protein Array
Classification Monoclonal
Clone CPTC-BCL2L2-2
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2
Host Species Mouse
Immunogen Recombinant human full-length BCL-W/BCL2L2 protein (Uniprot: Q92843)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 599
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.