Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bcl-x Antibody (5E9), Novus Biologicals™
SDP

Catalog No. NB298016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298016 50 μL
NB298017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298016 Supplier Novus Biologicals Supplier No. NBP32648050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

bcl-x Monoclonal antibody specifically detects bcl-x in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen bcl-x
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5E9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Apoptosis regulator Bcl-X,BCL2L,BCL2L1,Bcl-2-like protein 1,BCLX,Bcl-X,BCL-XL/S,PPP1R52
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Bcl-x [UniProt Q07817]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 598
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.